Skip to Content
Merck
All Photos(1)

Documents

794-0441

GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber

suitable for GC, application for SPME

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable
solid phase microextraction (SPME): suitable

test parameters

sample/matrix: 200 ppt each in 2 mL water
SPME fiber: polydimethylsiloxane, 100 μm (57300-U)
extraction: 15 min immersed in water with rapid stirring
column: SPB-5, 15 m × 0.20 mm I.D., 0.20 μm (24165-U)
oven: 120 °C (1 min) to 180 °C at 30 °C/min, then to 290 °C at 10 °C/min
inj. temp.: 260 °C (splitless - closed 3 min)
detector: ECD, 300 °C
carrier gas: helium, 37 cm/sec (set at 120 °C)

suitability

application for SPME

application(s)

environmental
food and beverages

Legal Information

SPB is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    4,4′-DDD, PESTANAL®, analytical standard
    Endrin aldehyde
    Aldrin
    Dieldrin
    α-BHC
    Endosulfan sulfate
    Endrin ketone
    α-Chlordane
    γ-Chlordane
    δ-BHC
    Decachlorobiphenyl
    4,4′-DDT
    Methoxychlor
    Heptachlor
    γ-BHC, certified reference material, 1000 μg/mL in methanol
    Endrin
    β-BHC
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene
    Heptachlor epoxide, PESTANAL®, analytical standard, isomer B
    See All (19)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service