Skip to Content
Merck
All Photos(10)

Documents

ZRB1776

Sigma-Aldrich

Anti-Syk Antibody, clone 2G22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Tyrosine-protein kinase SYK;EC:2.7.10.2;Spleen tyrosine kinase;p72-Syk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 72.07 kDa
observed mol wt ~70 kDa

purified by

using Protein A

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

canine, feline, porcine, bovine, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SYK(6850)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2G22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Syk. It targets an epitope within 16 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal half of human Tyrosine-protein kinase SYK (Syk).

Application

Quality Control Testing

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Syk in Ramos cell lysate.

Tested applications


Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Syk in lysates from A431 and Raw264.7 cells and Mouse Spleen tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Syk in human tonsil tissue sections.

Flow Cytometry Analysis: 1 μg from a representative lot detected Syk in one million Ramos cells.

Affinity Binding Assay: A representative lot of this antibody bound Syk with a KD of 6.6 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Tyrosine-protein kinase SYK (UniProt: P43405; also known as EC:2.7.10.2, Spleen tyrosine kinase, p72-Syk) is encoded by the SYK gene (Gene ID: 6850) in human. Syk is a non-receptor tyrosine kinase that is highly expressed in hematopoietic cells. It plays a crucial role not only in adaptive immune receptor signaling, but also in cellular adhesion, innate immune recognition, osteoclast maturation, platelet activation and vascular development. Syk contains two SH2 domains (aa 15-107 and 168-259) that mediate its interaction with the phosphorylated ITAM domains of transmembrane proteins. Its protein kinase domain is localized in amino acids 371-631. Intramolecular binding of the interdomains A and B (aa 108-160 and 260-370) to parts of its catalytic domain keeps the catalytic center in an inactive conformation. The phosphorylation of the interdomains or the binding of the SH2 domains with dually phosphorylated ITAM domains on transmembrane proteins disrupts these intramolecular interactions and allows the kinase domain to adopt an active conformation. Initial ITAM binding triggers rapid activation of Syk, whereas subsequent phosphorylation leads to prolonged activation. The phosphorylation of Syk and of the ITAM domains is essentially mediated by SRC subfamily kinases, like LYN, upon transmembrane receptors engagement. Syk can phosphorylate ITAMs and autophosphorylate its own interdomain region tyrosines, providing a positive feedback loop following initial ITAM-mediated Syk activation. Syk is also reported to mediate signaling of receptors that do not contain an ITAM, including integrins and C-type lectins, via interaction with ITAM-containing adaptors, such as DAP12 (TYROBP) and the FcR -chain (FcR ). This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Mocsai, A., et al. (2010). Nat. Rev. Immunol. 10(6); 387-402).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service