Z744274
Wilmad® Suprasil® EPR Tissue Sample Cell
well depth 0.5 mL, rod L 90 mm , upper, rod L 60 mm , lower
Synonym(s):
EPR sample cell, sample cell
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WG-806-A-Q
rod L
60 mm , lower
90 mm , upper
well depth
0.5 mL
Looking for similar products? Visit Product Comparison Guide
General description
EPR Tissue Sample Cells are used for viscous samples such as polymers and wet tissue. All cells include a 0.5 mm deep well for thick samples. The sample is placed between the plates. EPR Tissue Sample Cells come with two support rods. Each cell includes two coverslides and two “Bronzite” clips. Optional TPX plastic is highly permeable to O2, N2 and CO2 and should not be used with organic solvents.
Application
EPR
Legal Information
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service