Skip to Content
Merck
All Photos(1)

Documents

V9876

Sigma-Aldrich

cis-Vaccenoyl chloride

~98% (capillary GC), liquid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H33ClO
CAS Number:
Molecular Weight:
300.91
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~98% (capillary GC)

form

liquid

bp

170 °C/2 mmHg (lit.)

density

0.912 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCC\C=C/CCCCCCCCCC(Cl)=O

InChI

1S/C18H33ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h7-8H,2-6,9-17H2,1H3/b8-7-

InChI key

WYUXJEJFKYBBMZ-FPLPWBNLSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohei Kawabata et al.
Biological & pharmaceutical bulletin, 39(12), 1995-2008 (2016-12-03)
Different monounsaturated fatty acid (MUFA) species have distinct pathophysiological activities. cis-Palmitoleic acid (16:1n-7) was previously reported to improve insulin sensitivity in animal studies. The proportions of hepatic MUFAs are generally considered to reflect changes in the activities of fatty acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service