Documents
TMO312018-1000
Nalgene® square graduated bottle
high-density polyethylene, bottle capacity 1000 mL, natural, case of 250 ea
Synonym(s):
break resistant bottle, chemcial resistant bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
high-density polyethylene
natural
square
packaging
case of 250 ea
manufacturer/tradename
Nalgene 312018-1000
bottle capacity
1000 mL
General description
Save space with chemical- and break-resistant Thermo Scientific Nalgene Square HDPE Graduated Bottles with Closure. These translucent bottles provide visible liquid levels and molded-in graduations for easy measuring. Square bottles with polypropylene closures feature space-saving designs and drip-proof necks for convenient dispensing.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries