Skip to Content
Merck
All Photos(1)

Key Documents

T4678

Sigma-Aldrich

1,1,1-Trichloroethane

ACS reagent, 98.5-101.0% CH3CCl3 + active additives basis

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352103
NACRES:
NA.22

grade

ACS reagent

Assay

98.5-101.0% CH3CCl3 + active additives basis

contains

0.05% low alkyl epoxide as stabilizer

impurities

≤0.00030 meq/g Titratable acid
≤0.10% water

evapn. residue

≤0.001%

color

APHA: ≤10

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Ozone 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service