Skip to Content
Merck
All Photos(1)

Documents

T1264

Sigma-Aldrich

β-Thionaphthyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10OS
CAS Number:
Molecular Weight:
202.27
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

CC(=O)Sc1ccc2ccccc2c1

InChI

1S/C12H10OS/c1-9(13)14-12-7-6-10-4-2-3-5-11(10)8-12/h2-8H,1H3

InChI key

NMFMUXYHGNUOAI-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iu G Zhukovskiĭ et al.
Ukrains'kyi biokhimichnyi zhurnal (1999 ), 75(3), 67-70 (2003-10-28)
1- and 2-thionaphthylacetates were tested as cholinesterase substrates. It was shown that the butyrilcholinesterase from horse serum can hydrolize these compounds. The hydrolysis velocity of 1-thionaphthylacetate was comparable with hydrolysis velocity of acetylthiocholine (the well known cholinesterase substrate), but 2-thionaphthylacetate
B C Payne et al.
The Histochemical journal, 12(1), 71-86 (1980-01-01)
Using the substrate 2-naphthylthiol acetate (NTA), we developed a reproducible method of demonstrating a non-specific esterase while retaining nuclear and cytoplasmic details at the ultrastructural level. The NTA esterase had a distribution and pattern of staining similar to those of
H L Li et al.
Experimental and molecular pathology, 46(3), 321-330 (1987-06-01)
Nonspecific esterase activity was localized ultrastructurally within normal and leukemic hematopoietic cells by the use of 2-naphthylthiol acetate (NTA) as a substrate. NTA esterase activity was identified in all cell lines, although mononuclear phagocytes contained the most abundant activity. Monocytes
P L Sannes et al.
The Histochemical journal, 17(1), 43-56 (1985-01-01)
A 2-thionaphthyl acetate substrate was utilized to assess the subcellular distribution of nonspecific esterases in rat pulmonary alveolar and peritoneal macrophages. The enzymatically liberated 2-thionaphthol was visualized at pH 7.1 by utilizing gold as a capture agent. Glutaraldehyde-fixed macrophages derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service