Skip to Content
Merck
All Photos(1)

Documents

SML1347

Sigma-Aldrich

Alvimopan

≥98% (HPLC)

Synonym(s):

ADL-8-2698, LY-246736, N-[(2S)-2-[[(3R,4R)-4-(3-Hydroxyphenyl)-3,4-dimethyl-1-piperidinyl]methyl]-1-oxo-3-phenylpropyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32N2O4
CAS Number:
Molecular Weight:
424.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +46 to +52°, c = 1.0 in DMSO

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@H]1CN(C[C@@H](C(NCC(O)=O)=O)CC2=CC=CC=C2)CC[C@]1(C3=CC=CC(O)=C3)C

InChI

1S/C25H32N2O4/c1-18-16-27(12-11-25(18,2)21-9-6-10-22(28)14-21)17-20(24(31)26-15-23(29)30)13-19-7-4-3-5-8-19/h3-10,14,18,20,28H,11-13,15-17H2,1-2H3,(H,26,31)(H,29,30)/t18-,20-,25+/m0/s1

InChI key

UPNUIXSCZBYVBB-JVFUWBCBSA-N

Gene Information

human ... OPRM1(4988)

Biochem/physiol Actions

Alvimopan is a gastroprokinetic peripheral mu-opioid receptor antagonist. As a drug, Alvimopan is given to patients undergoing bowel surgery to protect the bowel from the effects of opioid medications that are used to treat pain, blocking the undesirable GI side-effects of the opioid agonists without affecting analgesia or causing withdrawal. Alvimopan does not cross the blood-brain barrier, competitively binding to gastrointestinal tract mu-opioid receptors.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service