Skip to Content
Merck
All Photos(1)

Documents

SAB1300583

Sigma-Aldrich

Anti-FUBP3 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FUBP3, Anti-FUSE-binding protein 3, Anti-Far upstream element-binding protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FUBP3(8939)

General description

The far upstream element-binding proteins FUBP, FUBP2, and FUBP3 comprise a family of single-strand DNA-binding proteins that possess all of the general features of more conventional transcription factors. The FUBPs each bind to a single sequence-specific strand of the far upstream element (FUSE; originally identified upstream of c-myc), and each possesses potent activation domains when fused to the GAL4 DNA-binding domain and assayed by transient transfection. These proteins have also been reported to bind RNA and participate in various steps of RNA processing, transport or catabolism.

Immunogen

FUBP3 (XP_001128545, 248-282)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the central region of human FUBP3.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weirui Ma et al.
Cell, 175(6), 1492-1506 (2018-11-20)
Approximately half of human genes generate mRNAs with alternative 3' untranslated regions (3'UTRs). Through 3'UTR-mediated protein-protein interactions, alternative 3'UTRs enable multi-functionality of proteins with identical amino acid sequence. While studying how information on protein features is transferred from 3'UTRs to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service