PQ0201
ProteoQwest™ Chemiluminescent Western Blotting Kit, CPS Substrate
for Mouse Monoclonal IgG Antibodies
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
antibody product type
secondary antibodies
storage temp.
2-8°C
General description
ProteoQwest Chemiluminescent Western Blotting Kit, CPS Substrate, includes essential reagents and antibodies used in a Western blot specific for mouse monoclonal IgG antibodies. This ProteoQwest kit is designed for high sensitivity chemiluminescent detection of as little as 0.3 ng of protein. The chemiluminescent reaction occurs directly on the protein immobilizing membrane. This kit is optimized for use with either film or chemiluminescent imagers. All components of the ProteoQwest Chemiluminescent Western Blotting Kit, CPS Substrate have been extensively tested and optimized to ensure superb results. This kit is designed for 12 mini-gel sized (10 × 10 cm) blots. Chemiluminescent Peroxidase Substrate (CPS-1), found in this kit, is a two component liquid substrate system able to detect picomole levels of protein with a superior signal to noise ratio.
Legal Information
ProteoQwest is a trademark of Sigma-Aldrich Co. LLC
Not finding the right product?
Try our Product Selector Tool.
Kit Components Only
Product No.
Description
- Rabbit anti-mouse IgG HRP-conjugated antibody 75 μL
- Chemiluminescent Reaction Buffer 80 mL
- Chemiluminescent Reagent 40 mL
Kit Components Also Available Separately
Product No.
Description
SDS
- Chemichrome™ Western Control 200 μL
- W0138Western Blocker™ Solution, for HRP detection systems 400 mLSDS
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bahareh Aliabedi et al.
Asian Pacific journal of cancer prevention : APJCP, 23(4), 1159-1167 (2022-04-30)
Growing evidence has demonstrated that microRNAs have a major effect on development of different types of cancer including AML. The overexpression of miR-625 could decrease tumorgenesis of acute myeloid leukemia cell lines through Integrin-linked kinase signaling pathway and reducing the
Mehdi Alikhani et al.
Iranian biomedical journal, 17(2), 54-61 (2013-04-10)
RNA-binding motif gene on Y chromosome (RBMY), a germ cell-specific nuclear protein, is known as a key factor in spermatogenesis and disorders associated with this protein have been recognized to be related to male infertility. Although it was suggested that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service