Skip to Content
Merck
All Photos(1)

Documents

P0896

Sigma-Aldrich

Diethyl pimelate

~97% (capillary GC)

Synonym(s):

Diethyl heptanedioate, Pimelic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)5COOC2H5
CAS Number:
Molecular Weight:
216.27
Beilstein:
1710126
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

~97% (capillary GC)

refractive index

n20/D 1.430 (lit.)

bp

192-194 °C/100 mmHg (lit.)

density

0.994 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)CCCCCC(=O)OCC

InChI

1S/C11H20O4/c1-3-14-10(12)8-6-5-7-9-11(13)15-4-2/h3-9H2,1-2H3

InChI key

LKKOGZVQGQUVHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service