Documents
M6414
Millipore® solid-phase extraction apparatus vacuum manifold
3-place kit for 90 mm filters
About This Item
UNSPSC Code:
41104900
manufacturer/tradename
Millipore XX0090SPE
General description
Dramatically reduces extraction times for environmental analysis. Analytes retained on a solid support can be eluted with a small quantity of solvent. Three-way valve allows sample filtration, washing, and elution without dismantling.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Kit Components Only
Product No.
Description
  • 90 mm anodized aluminum spring clamp 3 ea
  • 90 mm base with 3-way valve 3 ea
  • 1-liter glass funnel 3 ea
  • 90 mm PTFE-coated, stainless steel screen 3 ea