Skip to Content
Merck
All Photos(4)

Documents

HPA042981

Sigma-Aldrich

Anti-NLRP12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Clr19.3, Anti-Monarch1, Anti-Nalp12, Anti-Nlr family, pyrin domain containing 12, Anti-Pan6, Anti-Pypaf7, Anti-Rno2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LKRCRSAQVLHLYGATYSADGEDRARCSAGAHTLLVQLPERTVLLDAYSEHLAAALCTNPNLIELSLYRNALGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NLRP12(91662)

General description

NLR family pyrin domain containing 12 (NLRP12), also known as PYRIN-containing Apaf1-like protein 7 (PYPAF7), is a novel PYPAF protein and is a member of CATERPILLER protein family. It is encoded by the gene mapped to human chromosome 19q13.4. The protein is specifically expressed in immune cells.

Immunogen

NLR family, pyrin domain containing 12 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NLR family pyrin domain containing 12 (NLRP12) plays a vital role in regulation of inflammatory signaling pathways by controlling the activation of NF-ΚB and cytokine processing. Variation in the gene expression leads to hereditary periodic fever syndromes. NLRP12 regulates intestinal homeostasis and delivers protection against colorectal tumorigenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82718

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PYPAF7, a novel PYRIN-containing Apaf1-like protein that regulates activation of NF-kappa B and caspase-1-dependent cytokine processing.
Wang L
The Journal of Biological Chemistry null
Nidhi Sharma et al.
Scientific reports, 9(1), 8480-8480 (2019-06-13)
Gliomas are the most prevalent primary brain tumors with immense clinical heterogeneity, poor prognosis and survival. The nucleotide-binding domain, and leucine-rich repeat containing receptors (NLRs) and absent-in-melanoma 2 (AIM2) are innate immune receptors crucial for initiation and progression of several
Valentina Galbiati et al.
Toxicology letters, 313, 130-136 (2019-07-06)
We previously demonstrated that based on their potency, contact allergens differently modulate Blimp-1/NLRP12 expression in human keratinocytes, with the extreme allergen 2,4-dinitrochlorobenzene (DNCB) more rapidly upregulating Blimp-1, leading to downregulation of NLRP12, and to the production of interleukin-18 (IL-18). The
Mutations in NALP12 cause hereditary periodic fever syndromes.
Jeru I
Proceedings of the National Academy of Sciences of the USA null
The NOD-like receptor NLRP12 attenuates colon inflammation and tumorigenesis.
Zaki MH
Cancer Cell null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service