OX-42 Cell Line from murine myeloma
Reactivity: Anti- CD11, rat, 87081803
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
NS0/U
Reactivity
Anti-CD11, rat
technique(s)
cell culture | mammalian: suitable
isotype
IgG2a
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse NS0/U x Mouse BALB/c spleen
Cell Line Description
The antibody detects rat CD11. All peritoneal macrophages and 35% of alveolar macrophages react. The antibody affects the function of complement receptor CR3. It immunoprecipitates 3 polypeptides: 160kDa, 130kDa, 95kDa. The antibody also reacts with granulocytes and dendritic cells.
Immunogen
Resident peritoneal cells from (PVG-RT1c x PVG RT1u)F1 and (PVG-RT1c x DA-RT1a)F1 rats
Culture Medium
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Seed at 5x10,000 viable cells/ml. Subculture when cells reach 5x100,000-1x1,000,000 cells/ml (usually after 3-4 days); 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service