C4726
EX-CELL® CD CHO-2 Medium
Animal-component free, with sodium bicarbonate, without L-glutamine, without phenol red., liquid, sterile-filtered, suitable for cell culture
Synonym(s):
CHO Medium
About This Item
description
for research or for further manufacturing use
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
components
phenol red: no
HEPES: yes
NaHCO3: yes
L-glutamine: no
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
Other Notes
Does not contain antibiotics, antimycotics, L-glutamine, transferrin, hydrolysates, or other undefined nutrients or supplements. Contains no animal-derived components or components synthesized from animal-derived materials.
Reconstitution
Legal Information
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service