Skip to Content
Merck
All Photos(1)

Documents

93359

Sigma-Aldrich

TES

BioUltra, ≥99.5% (calc. based on dry substance, T)

Synonym(s):

2-[(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]ethanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid, TES free acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO6S
CAS Number:
Molecular Weight:
229.25
Beilstein:
1957061
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Assay

≥99.5% (calc. based on dry substance, T)

form

powder

impurities

insoluble matter, passes filter test

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying, 110 °C

pH

3.5-5.0 (25 °C, 1 M in H2O)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

mp

~225 °C (dec.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.055
λ: 280 nm Amax: 0.045

SMILES string

OCC(CO)(CO)NCCS(O)(=O)=O

InChI

1S/C6H15NO6S/c8-3-6(4-9,5-10)7-1-2-14(11,12)13/h7-10H,1-5H2,(H,11,12,13)

InChI key

JOCBASBOOFNAJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TES is a physiological Good′s buffer (pH 74.-7.9) used in biological research applications. It is often compared to other Good′s buffers for chemistry and optimal performance in a given system.

Other Notes

Hydrogen ion buffer for biological research; Suitable buffer for epidermal cell growth at pH 7.4-7.9 where additional buffering capacity is required

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Hammar et al.
Epidermis Dis., Proc. Eur. Soc. Dermatol. Res. Symp. 1979, 243-243 (1981)
Yikang Wu et al.
Organic letters, 4(13), 2141-2144 (2002-06-21)
[reaction: see text] In DMSO cleavage of triethylsilyl (TES) ethers by o-iodoxybenzoic acid (IBX) was significantly faster than cleavage of tert-butyldimethylsilyl (TBS) ethers or further oxidation into carbonyl compounds. In most cases, TES protecting groups could be removed in good
M J Taylor et al.
The British journal of ophthalmology, 73(10), 781-791 (1989-10-01)
Preservation solutions for short-term storage of isolated donor corneas for use in penetrating keratoplasty have all been based on tissue culture medium, on the assumption that media designed to maintain the viability of cells at physiological temperatures will also provide
R H Martin et al.
Biology of reproduction, 47(2), 268-270 (1992-08-01)
Human sperm karyotypes can be prepared after fusion of human sperm with Golden hamster oocytes. Most laboratories use one of two methods of sperm capacitation: incubation of freshly-ejaculated sperm in Biggers, Whitten, and Whittingham (BWW) medium for 5-7 h at
N Sugiyama et al.
Molecular reproduction and development, 58(1), 30-38 (2001-01-06)
The growth, differentiation, and death/survival of spermatogonia are precisely regulated for the proper production of spermatozoa. We have previously shown that Bcl-2 ectopically expressed in spermatogonia caused the inhibition of normal spermatogonial apoptosis and the subsequent failure of differentiation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service