71723
Sodium DL-lactate solution
50% in H2O
Synonym(s):
DL-Lactic acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solution
concentration
50% in H2O
SMILES string
[Na+].CC(O)C([O-])=O
InChI
1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1
InChI key
NGSFWBMYFKHRBD-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
The product is a 50% solution of sodium DL-lactate in H2O. Sodium DL-lactate is the sodium salt of DL-lactic acid.
Application
Sodium DL-lactate (DL-lactate) may be employed as a carbon source in the minimal salt medium (MSM) for the culture of Pseudomonas putida KT2440. It may be employed as an energy source in the basal complex medium for the growth of Selenomonas ruminantium strains.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Applied microbiology, 30(6), 916-921 (1975-12-01)
A lactate-fermenting strain of Selenomonas ruminantium (HD4) and a lactatenonfermenting strain (GA192) were examined with respect to the stereoisomers of lactate formed during glucose fermentation, the stereoisomers of lactate fermented by HD4, and the characteristics of the lactate dehydrogenases of
Scientific reports, 4, 6939-6939 (2014-11-07)
As an important method for building blocks synthesis, whole cell biocatalysis is hindered by some shortcomings such as unpredictability of reactions, utilization of opportunistic pathogen, and side reactions. Due to its biological and extensively studied genetic background, Pseudomonas putida KT2440
Critical care (London, England), 18(2), R48-R48 (2014-03-29)
Acute heart failure (AHF) is characterized by inadequate cardiac output (CO), congestive symptoms, poor peripheral perfusion and end-organ dysfunction. Treatment often includes a combination of diuretics, oxygen, positive pressure ventilation, inotropes and vasodilators or vasopressors. Lactate is a marker of
Influence of lactate and acetate salt adaptation on Salmonella Typhimurium acid and heat resistance.
Food microbiology, 30(2), 448-452 (2012-03-01)
The aim of the present study was to determine the survival of Salmonella Typhimurium adapted with sodium lactate (NaL), potassium lactate/sodium acetate mixture (KL/NaA) or sodium acetate (NaA) in simulated gastric fluid (SGF) and during heat treatment. NaL-, KL/NaA- and
Transplantation, 97(12), 1272-1278 (2014-03-22)
Over 40% of deceased donors in the UK donate after circulatory death (DCD). Normothermic regional perfusion has been reported to improve outcomes in such donors in Europe and the United States. Unlike the United States, legal and professional requirements in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service