Skip to Content
Merck
All Photos(1)

Key Documents

11024

Sigma-Aldrich

Aluminum nitrate nonahydrate

technical, ≥98%

Synonym(s):

Aluminium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
375.13
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

technical

Assay

≥98%

impurities

≤0.01% heavy metals (as Pb)

mp

73 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Fe: ≤100 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Al.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SWCIQHXIXUMHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service