PRIH3239D0B2
Princeton BioMolecular Research, Inc.
2-Cycloheptyl-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
287.32
MDL number:
Iupac Name
2-cycloheptyl-1,3-dioxo-5-isoindolinecarboxylic acid
Inchi Code
1S/C16H17NO4/c18-14-12-8-7-10(16(20)21)9-13(12)15(19)17(14)11-5-3-1-2-4-6-11/h7-9,11H,1-6H2,(H,20,21)
InChI key
NMKHNBKGJXJOHW-UHFFFAOYSA-N
Purity
95%
Country of Origin
UA
Physical Form
Solid
Product Link
http://store.princetonbio.com/?product_id=PBMR057921
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.