Skip to Content
Merck
All Photos(3)

Documents

RDYTUBE10

Millipore

ReadyStream® Inlet Tube

Tubing for connection of ReadyStream® system to a water source

Synonym(s):

Inlet Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102014
NACRES:
NB.11

material

acetal (polypropylene)

packaging

pkg of 1 unit

system L × W × H

21 cm × 10 cm × 5 cm

color

white

application(s)

food and beverages

compatibility

for use with ReadyStream® System

storage temp.

15-25°C

General description

ReadyStream® Inlet Tube is designed to connect the ReadyStream® instrument to a water source in your lab. It is also possible to put the tubing inside a canister with water suitable for ReadyStream® from where the water can be pumped.

Application

ReadyStream® Inlet Tube is an accessory of the ReadyStream® system used to connect the instrument to a water source in the lab.

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service