MABN834
Anti-SOD1 Antibody (ALS mutant), clone MS785
clone MS785, from rat
Synonym(s):
Superoxide dismutase [Cu-Zn], Superoxide dismutase 1, hSod1, SOD1
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
MS785, monoclonal
species reactivity
human
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SOD1(6647)
General description
Immunogen
Application
and Immunoprecipitation.
Western Blotting Analysis: A representative lot SOD1 in transfected HEK293 cells (Fujisawa, T., et al. (2012). Ann Neurol. 72: 739–749).
Immunoprecipitation Analysis: A representative lot immunoprecipitated SOD1 in transfected HEK293 cells (Homma, K., et al. (2013). Ann Neurol. Cell. 52:1-12).
Immunocytochemistry (ICC): A representative lot of this antibody detected SOD1-G93A in HEK293 cells overexpressing G93A, but not the wild-type SOD1. (Fujisawa, T., et al. 2015. Neurobiol. Dis. 82; 486-487).
Immunohistochemistry (IHC): A representative lot of this antibody detected SOD1-G93A in the anterior horns of spinal cord tissue sections of SOD1G93A-transgenic mice, but not non-transgenic mice. ((Fujisawa, T., et al. 2015. Neurobiol. Dis. 82; 486-487).
Neuroscience
Developmental Signaling
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected SOD1 in 1 µg of transfected HEK293 with SOD1 mutant and HEK293 control lysates.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service