Skip to Content
Merck
All Photos(3)

Documents

MAB413-I

Sigma-Aldrich

Anti-Interferon-α Antibody

purified antibody, clone ST126, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ST126, monoclonal

mol wt

calculated mol wt 21.55 kDa
observed mol wt ~19 kDa

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
neutralization: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Specificity

Clone ST126 is a mouse monoclonal antibody that detects human Interferon a.

Immunogen

A full-length purified human interferon-a produced by human Burkitt lymphoma Namalwa cell line.

Application

Quality Control TestingEvaluated by Western Blotting with recombinant human IFN- a2.Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant human IFN-a2.Tested ApplicationsNeutralizing: A representative lot of this antibody neutralized the antiviral activity of Interferon-a. (Shearer, M., et. al. (1984). J Immunol. 133(6):3096-101).ELISA Analysis: A representative lot detected Interferon-a in ELISA applications (Shearer, M., et. al. (1984). J Immunol. 133(6):3096-101).Radioimmunoassay: A representative lot detected Interferon-a in Radioimmunoassay applications (Shearer, M., et. al. (1984). J Immunol. 133(6):3096-101). Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Interferon-a in human kidney and human stomach cancer tissue sections.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Interferon-a, clone ST126, Cat. No. MAB413-I, is a mouse monoclonal antibody that detects Interferon alpha-2 and is tested for use in ELISA, Immunohistochemistry (Paraffin), Neutralizing, Radioimmunoassay, and Western Blotting.

Target description

Interferon alpha-2 (UniProt: P01563; also known as IFN-alpha-2, Interferon alpha-A, LeIF A) is encoded by the IFNA2 (also known as IFNA2A, IFNA2B, IFNA2C) gene (Gene ID: 3440) in human. Interferons are divided into three families (type I, II, and III) based on their sequence homology. Type I is the largest group that includes Interferons a and b are the primary defensive cytokines against pathogens and display strong antiviral activities. They also display anti-bacterial and anti-cancer properties. IFN-a is rapidly induced and secreted by cells to limit virus replication through numerous and varied mechanisms, including the induction of intrinsic restriction factors. IFN-a2 is produced by macrophages with a signal peptide (aa 1-23), which is subsequently cleaved off to generate the mature active form. It is produced in response to viral infection and triggers the induction of protective genes that inhibit viral replication and impede viral dissemination. IFN-a2 is shown to be an effective agent against hepatitis viruses. Chronic administration of IFN-a is shown to cause the regression of vascular neoplasms, including hemangiomas, Kaposi′s sarcoma, and pulmonary hemangiomatosis. (Ref.: Lavender, KJ., et al. (2016). J, Virol. 90(13); 6001-6013).

Physical form

Purified mouse monoclonal antibody IgG2b in PBS without azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service