MAB10209-I
Anti-HSV-2 Nucleocapsid p40 Antibody, clone 3E1
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
3E1, monoclonal
mol wt
calculated mol wt 66.94 kDa
purified by
using protein G
species reactivity
virus
packaging
antibody small pack of 100 μL
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
isotype
IgG1
epitope sequence
Unknown
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
General description
Specificity
Immunogen
Application
Evaluated by Immunohistochemistry (Paraffin) in HSV-2-positive tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected HSV-2 Nucleocapsid p40 in HSV-2-positive tissue sections.
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service