Skip to Content
Merck
All Photos(1)

Documents

ABF189

Sigma-Aldrich

Anti-FcRn/FCGRT Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

IgG receptor FcRn large subunit p51, FcRn, IgG Fc fragment receptor transporter alpha chain, Neonatal Fc receptor, FcRn/FCGRT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology), feline (based on 100% sequence homology), monkey (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FCGRT(2217)

General description

IgG receptor FcRn large subunit p51 (UniProt P55899; also known as FcRn, IgG Fc fragment receptor transporter alpha chain, Neonatal Fc receptor) is encoded by the FCGRT gene (also known as FCRN) (Gene ID 2217) in human. Transfer of passive immunity from mother to the fetus or newborn involves the transport of IgG across several epithelia. Depending on the species, IgG is transported prenatally across the placenta and yolk sac or is absorbed from colostrum and milk by the small intestine of the suckling newborn. In both cases, apical to basolateral transepithelial transport of IgG is mediated by FcRn. FcRn is a heterodimer of an alpha-chain and beta(2)-microglobulin (beta(2)m) and differs from other IgG Fc receptors in that it is structurally related to MHC class I molecules. The assembly of the FcRn alpha-chain with beta(2)m is important for both cell surface expression and pH-dependent high affinity IgG binding. Proteins endocytosed from serum are degraded in the lysosomes. However, serum albumin (SA) and IgG bind to FcRn at low pH in the endosome after endocytosis, and are transported back to the cellular surface, where they are released into the bloodstream, resulting in an extended serum half-life.

Immunogen

Epitope: Alpha-2/-3 domain
Linear peptide corresponding to the Alpha-2/-3 domain of human FcRn/FCGRT.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-FcRn/FCGRT Antibody is validated for use in Western Blotting for the detection of FcRn/FCGRT .
Western Blotting Analysis: A representative lot detected FcRn/FCGRT in FO-1 cells expressing Myc-hFcRn (Praetor, A., et al. (2002). J Cell Sci. 115(Pt11):2389-2397).
Western Blotting Analysis: A representative lot detected FcRn/FCGRT in hFcRn transfected MDCK cells (Praetor, A., et al. (1999). J Cell Sci. 112(Pt14):2291-2299).

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected FcRn/FCGRT in 10 µg of human liver tissue lysate.

Target description

~35/45 kDa observed, with the lower band representing non- and/or less glycosylated form(s).

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service