Skip to Content
Merck
All Photos(3)

Key Documents

ABE1367

Sigma-Aldrich

Anti-LCoR (Splice Variant)

from rabbit

Synonym(s):

Ligand-dependent corepressor, Mblk1-related protein 2, C10ORF12, LCoR (splice variant), PALI1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... LCOR(84458)

General description

Ligand-dependent corepressor (UniProt: Q96JN0; also known as LCoR, Mblk1-related protein 2) is encoded by the LCOR (also known as C10orf12, KIAA1795, MLR2) gene (Gene ID: 84458) in human. LCoR is a ubiquitously distributed protein in fetal and adult tissues and serves as a transcriptional co-repressor that is recruited to agonist-bound nuclear receptors through a single LxxLL motif at the N-terminal region. It may also act as a transcription activator that binds DNA elements with the sequence 5′-CCCTATCGATCGATCTCTACCT-3′. Three isoforms of LCoR have been described that are produced by alternative splicing. This polyclonal antibody is raised against a large isoform (isoform 3; a splice variant), which associates with the PRC2 complex through its C-terminal region. Its nuclear localization signal is reported to be in amino acids 339-345 and it reacts with nuclear receptors via amino acids 53-57. (Ref.: Conway, E., et al. (2018). Mol. Cell 70(3); 408-421).

Specificity

This rabbit polyclonal antibody detects a splice variant of human and murine Ligand-dependent corepressor (LCoR). It targets an epitope with in the N-terminal half.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal half of human Ligand-dependent corepressor (LCoR), isoform 3.

Application

Anti-LCoR, Cat. No. ABE1367, is a highly specific rabbit polyclonal antibody that targets a splice variant form of Ligand-dependent corepressor (LCoR) and has been tested in Immunocytochemistry and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:500 dilution from a representative lot detected LCoR in whole cell lysates from Pali1 KO and matched wild type mouse embryonic stem (Courtesy of Dr. Eric Conway, Dr. Emilia Jerman & Prof. Adrian P. Bracken at the Smurfit Institute of Genetics, Trinity College Dublin, Ireland).

Western Blotting Analysis: A representative lot detected LCoR in Western Blotting applications (Conway, E., et. al. (2018). Mol. Cell. 70(3):408-421).

Quality

Evaluated by Immunocytochemistry in NTERA2 cells.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected LCoR in NTERA2 cells.

Target description

~250 kDa observed; 170.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Zhang et al.
Nature communications, 12(1), 4592-4592 (2021-07-30)
The polycomb repressive complex 2 (PRC2) is a histone methyltransferase that maintains cell identities. JARID2 is the only accessory subunit of PRC2 that known to trigger an allosteric activation of methyltransferase. Yet, this mechanism cannot be generalised to all PRC2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service