Skip to Content
Merck
All Photos(1)

Documents

8.41828

Sigma-Aldrich

3-(Dimethylamino)-propiophenone hydrochloride

for synthesis

Synonym(s):

3-(Dimethylamino)-propiophenone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
213.7
MDL number:
UNSPSC Code:
12352107
EC Index Number:
212-909-1
NACRES:
NA.22

Quality Level

form

solid

mp

155-156 °C

storage temp.

2-30°C

InChI

1S/C11H15NO.ClH/c1-12(2)9-8-11(13)10-6-4-3-5-7-10;/h3-7H,8-9H2,1-2H3;1H

InChI key

DKNDBIIKSJWQFL-UHFFFAOYSA-N

General description

for synthesis

Application

3-(Dimethylamino)-propiophenone hydrochloride for synthesis. CAS 879-72-1, molar mass 213.7 g/mol.

Analysis Note

Assay (argentometric)≥ 99.0 %Identity (IR)passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service