Skip to Content
Merck
All Photos(1)

Key Documents

6.18628

Sigma-Aldrich

Methyl paraben (Methyl-4-hydroxybenzoate)

extra pure

Synonym(s):

Methyl 4-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(OH)C6H4(COOCH3)
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352100

grade

extra pure

InChI

1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3

InChI key

LXCFILQKKLGQFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service