Skip to Content
Merck
All Photos(1)

Key Documents

555556

Sigma-Aldrich

Rho Kinase Inhibitor VII

The Rho Kinase Inhibitor VII controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Rho Kinase Inhibitor VII, ROCK Inhibitor VII, Rho Kinase Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N8
Molecular Weight:
388.47
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light beige

solubility

DMSO: 1 mg/mL
water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable triaminotriazine ROCK inhibitor (IC50 = 6 nM in a cell-free ROCK1 kinase assay) that affects 24 other commonly studied non-ROCK kinases only at much higher concentrations (IC50 >2 µM) and effectively blocks MCP-1-induced THP-1 cell migration (IC50 = 30 nM) in vitro. Subcutaneous injection (3-10 mg/kg) of ROCK Inhibitor VII and oral dosing (30 mg/kg) of Y-27632 (Cat. Nos. 688000 & 688001) are both reported to significantly reduce arterial blood pressure in a spontaneous hypertensive rat model in vivo.
A cell-permeable triaminotriazine ROCK inhibitor (IC50 = 6 nM in a cell-free ROCK1 kinase assay) that affects 24 other commonly studied non-ROCK kinases only at much higher concentrations (IC50 >2 µM) and effectively blocks MCP-1-induced THP-1 cell migration (IC50 = 30 nM) in vitro. Subcutaneous injection (3-10 mg/kg) of ROCK Inhibitor VII and oral dosing (30 mg/kg) of Y-27632 (Cat. Nos. 688000 & 688001) are both reported to significantly reduce arterial blood pressure in a spontaneous hypertensive rat model in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ho, K.K., et al. 2009. Bioorg. Med. Chem. Lett.19, 6027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service