Skip to Content
Merck
All Photos(1)

Documents

400122-M

Sigma-Aldrich

Human IgG₂, Myeloma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

antibody form

purified immunoglobulin

Quality Level

Assay

>95% (SDS-PAGE)

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage temp.

−20°C

target post-translational modification

unmodified

General description

Native IgG2 from human myeloma plasma.
Native IgG2 from human myeloma plasma. Human IgG2 possessing a kappa (κ) light chain. Purified from human myeloma plasma.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 20 mM phosphate buffer, pH 7.4.

Preparation Note

Prepared from plasma that was shown by certified tests to be nonreactive for HBsAg and negative for antibodies to HCV, HBc, HIV-1, and HIV-2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Single arc by IEP.

Other Notes

The heavy chain of myeloma IgGs may appear as either a single or double band on gel electrophoresis. Prepared from plasma that was shown by certified tests to be nonreactive for HBsAg and negative for antibodies to HCV, HBc, HIV-1, and HIV-2. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service