Skip to Content
Merck
All Photos(1)

Documents

JWPH3249C55F

J & W Pharmlab LLC

2-Hydroxy-6,7,8,9-tetrahydro-benzocyclohepten-5-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176.22
MDL number:

Iupac Name

2-hydroxy-6,7,8,9-tetrahydro-5H-benzo[a]cyclohepten-5-one

Inchi Code

1S/C11H12O2/c12-9-5-6-10-8(7-9)3-1-2-4-11(10)13/h5-7,12H,1-4H2

InChI key

LCEJOJCDTXCOCR-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Off White Solid

Product Link

https://www.jwpharmlab.com/home/product/10R0688

MSDS

https://www.jwpharmlab.com/home/product/10R0688


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service