Skip to Content
Merck
All Photos(1)

Key Documents

ENAH90823717

Enamine

3-(2-methylpropyl)azetidin-3-ol; trifluoroacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$32.20
250 MG
$46.00
1 G
$115.00

Vendor SKU: D63461-100MG

$32.20


Ships in 10 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
100 MG
$32.20
250 MG
$46.00
1 G
$115.00

About This Item

CAS Number:
Molecular Weight:
243.23
MDL number:

Vendor SKU: D63461-100MG

$32.20


Ships in 10 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

3-isobutylazetidin-3-ol 2,2,2-trifluoroacetate

Inchi Code

1S/C7H15NO.C2HF3O2/c1-6(2)3-7(9)4-8-5-7;3-2(4,5)1(6)7/h6,8-9H,3-5H2,1-2H3;(H,6,7)

InChI key

AMRUUJCUBVCNPT-UHFFFAOYSA-N

storage temp.

4

Salt Data

OC(=O)C(F)(F)F

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

oil

Product Link

https://www.enaminestore.com/catalog/EN300-7540322

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-7540322


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library