ENAH2C55703D
Enamine
tert-butyl 4-(3,3-difluorocycloprop-1-en-1-yl)piperidine-1-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl 4-(3,3-difluorocycloprop-1-en-1-yl)piperidine-1-carboxylate
Inchi Code
1S/C13H19F2NO2/c1-12(2,3)18-11(17)16-6-4-9(5-7-16)10-8-13(10,14)15/h8-9H,4-7H2,1-3H3
InChI key
QDPJDTCUSCCERA-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-1722269
MSDS
https://www.enaminestore.com/catalog/EN300-1722269
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.