Skip to Content
Merck
All Photos(1)

Key Documents

ENA517162565

Enamine

methyl 5-(chlorosulfonyl)naphthalene-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
284.72
MDL number:

mp

114-115

Iupac Name

methyl 5-(chlorosulfonyl)-1-naphthoate

Inchi Code

1S/C12H9ClO4S/c1-17-12(14)10-6-2-5-9-8(10)4-3-7-11(9)18(13,15)16/h2-7H,1H3

InChI key

CNFVMBKEYPEFQX-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-312848

Melting Point

114-115

MSDS

https://www.enaminestore.com/catalog/EN300-312848


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library