ENA371177975
Enamine
1,3,3-trimethylbicyclo[2.2.1]heptane-2-carbonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1,3,3-trimethylbicyclo[2.2.1]heptane-2-carbonitrile
Inchi Code
1S/C11H17N/c1-10(2)8-4-5-11(3,6-8)9(10)7-12/h8-9H,4-6H2,1-3H3
InChI key
OHWRGMHXQWAJAZ-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-105779
MSDS
https://www.enaminestore.com/catalog/EN300-105779
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.