COM448619487
Combi-Blocks, Inc.
4-(3-(3-Bromophenyl)propyl)morpholine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-[3-(3-bromophenyl)propyl]morpholine
Inchi Code
1S/C13H18BrNO/c14-13-5-1-3-12(11-13)4-2-6-15-7-9-16-10-8-15/h1,3,5,11H,2,4,6-10H2
InChI key
BRDFMYRGMMWEOR-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-2661
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-2661
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.