Skip to Content
Merck
All Photos(1)

Documents

CIAH98DE3858

ChemScene LLC (Preferred Partner)

1,2,3,4-Tetrahydroquinolin-8-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
148.21
MDL number:

bp

145 C at 760 mmHg

Iupac Name

1,2,3,4-tetrahydro-8-quinolinamine

Inchi Code

1S/C9H12N2/c10-8-5-1-3-7-4-2-6-11-9(7)8/h1,3,5,11H,2,4,6,10H2

InChI key

LILSBXJJIIFDGR-UHFFFAOYSA-N

storage temp.

4C, protect from light

Purity

95%

Country of Origin

CN

Physical Form

Viscous Liquid

Product Link

https://www.chemscene.com/54012-92-9.html

Boiling point

145 C at 760 mmHg

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0314714.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service