ATE939904710
AstaTech, Inc.
METHYL 6-(1-AMINOCYCLOPROPYL)NICOTINATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 6-(1-aminocyclopropyl)nicotinate
Inchi Code
1S/C10H12N2O2/c1-14-9(13)7-2-3-8(12-6-7)10(11)4-5-10/h2-3,6H,4-5,11H2,1H3
InChI key
QAWLOHWWIRGDCJ-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Physical Form
Pale-yellow to Yellow-brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=107498
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=95054
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.