Skip to Content
Merck
All Photos(1)

Key Documents

ATE428009971

AstaTech, Inc.

TERT-BUTYL 4-(2-HYDROXY-4-METHYL PHENYLAMINO)PIPERIDINE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
306.41
MDL number:

Iupac Name

tert-butyl 4-(2-hydroxy-4-methylanilino)-1-piperidinecarboxylate

Inchi Code

1S/C17H26N2O3/c1-12-5-6-14(15(20)11-12)18-13-7-9-19(10-8-13)16(21)22-17(2,3)4/h5-6,11,13,18,20H,7-10H2,1-4H3

InChI key

XJSQGAFXUVITMJ-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=39963

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AB6695


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library