AMBH2D6ECC5B
Ambeed, Inc.
6-Bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydro-naphthalene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
6-bromo-1,1,4,4,7-pentamethyl-1,2,3,4-tetrahydronaphthalene
Inchi Code
1S/C15H21Br/c1-10-8-11-12(9-13(10)16)15(4,5)7-6-14(11,2)3/h8-9H,6-7H2,1-5H3
InChI key
ONNHBALCPUEXBT-UHFFFAOYSA-N
storage temp.
Sealed in dry,2-8C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/119999-22-3.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.