Z342157
IKA® multi position hotplate stirrer
No. of stirring stations: 10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41102406
Recommended Products
AC/DC input
230 V
230 V
material
silicone plate
feature
CE compliant
manufacturer/tradename
RT 10 power
IKA 2930500
availability
not available in USA and Canada
parameter
≤100 °C temperature
0-1200 rpm speed
120-248 °C temp. range
plate size × W × L
× 178 mm × 452 mm
Looking for similar products? Visit Product Comparison Guide
General description
The RT series is a high-performance multi-station magnetic stirrer with integrated temperature control plate. Precise temperature distribution on the heating plate makes it possible to perform serial experiments.
The synchronously running stirring stations also ensure that the conditions of the experiment will be completely comparable for the individual samples.
Please note: Water bath attachments are no longer available for this particular range of hotplate stirrers (5, 10 and 15 - stirring positions).
The synchronously running stirring stations also ensure that the conditions of the experiment will be completely comparable for the individual samples.
Please note: Water bath attachments are no longer available for this particular range of hotplate stirrers (5, 10 and 15 - stirring positions).
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service