Skip to Content
Merck
All Photos(1)

Key Documents

W325406

Sigma-Aldrich

(+)-Arabinogalactan

Synonym(s):

Polyarabinogalactan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3254
EC Number:
MDL number:
UNSPSC Code:
12164502

biological source

synthetic

optical activity

[α]25/D +11.5°, c = 1 in H2O

mp

>200 °C (dec.) (lit.)

application(s)

flavors and fragrances

Organoleptic

pine

InChI

1S/C20H36O14/c1-7-11(23)18(12(24)9(4-21)32-7)34-20-15(27)17(29-3)13(25)10(33-20)6-31-19-14(26)16(28-2)8(22)5-30-19/h7-27H,4-6H2,1-3H3

InChI key

SATHPVQTSSUFFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dietary fiber that enhances immune system function by stimulating natural killer (NK) cell activity.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Lluis et al.
Current opinion in allergy and clinical immunology, 12(2), 158-163 (2012-02-07)
Several population-based studies have replicated the finding that exposure to a farm environment is protective against the development of atopic diseases. From these studies, novel insights into potential allergy-protective mechanisms were retrieved. This review focuses on consistent and novel findings
A Passariello et al.
Alimentary pharmacology & therapeutics, 35(7), 782-788 (2012-02-14)
Acute diarrhoea is a frequent problem in children with heavy economic burden for families and society. To test the efficacy of a new synbiotic formulation containing Lactobacillus paracasei B21060, arabinogalactan and xilooligosaccharides in children with acute diarrhoea. Double-blind, randomised, placebo-controlled
Izabela Pawlaczyk et al.
Carbohydrate polymers, 92(1), 741-750 (2012-12-12)
From the air-dried Wild strawberry (Fragaria vesca L., family Rosaceae) leaves five water-soluble glycoconjugates Fv I-V by different extraction conditions have been isolated. Effects of extraction steps/agents on chemical composition and anticoagulant activity of Fv I-V were examined. Dark brown
Andreas Herrmann et al.
Glycobiology, 22(11), 1424-1439 (2012-07-10)
Water-soluble high molecular weight compounds were isolated in yields of 21-22% from the oleogum of Boswellia serrata and B. carteri. Using anion exchange chromatography and gel permeation chromatography, different proteoglycans were purified and characterized, leading to four principally different groups:
Gérald Larrouy-Maumus et al.
The Journal of biological chemistry, 287(47), 39933-39941 (2012-10-06)
The biosynthesis of the major cell envelope glycoconjugates of Mycobacterium tuberculosis is topologically split across the plasma membrane, yet nothing is known of the transporters required for the translocation of lipid-linked sugar donors and oligosaccharide intermediates from the cytoplasmic to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service