THS03148
ThalesNano MicroCatCart™ catalyst cartridge system
L × diam. 30 mm × 4 mm, 1% Pd/Al2O3
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 6 ea
manufacturer/tradename
ThalesNano THS 03148
L × diam.
30 mm × 4 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
MicroCatCart is a trademark of ThalesNano Nanotechnology Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service