BR94905
BRAND® standard ground joint funnel
funnel O.D. 40 mm
Synonym(s):
Funnels, Standard ground joint funnels
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Recommended Products
material
polypropylene
Joint
joint ST/NS14/23
packaging
pack of 10 ea
manufacturer/tradename
BRAND 94905
funnel O.D.
40 mm
Related Categories
General description
Half-round body. Stem fits standard ground sockets. Ideal for filling liquids or powders into multi-neck flasks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)