Skip to Content
Merck
All Photos(1)

Documents

A76907

Sigma-Aldrich

6-Thioguanine

98%

Synonym(s):

2-Amino-6-mercaptopurine, 2-Amino-6-purinethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5S
CAS Number:
Molecular Weight:
167.19
Beilstein:
157765
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

98%

mp

≥300 °C (lit.)

SMILES string

NC1=Nc2nc[nH]c2C(=S)N1

InChI

1S/C5H5N5S/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)

InChI key

WYWHKKSPHMUBEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ribosylated and phosphorylated by the same pathway as natural purine bases; as the nucleotide, inhibits a variety of cellular processes involved in nucleic acid synthesis. Has a long history as an effective treatment of leukemia.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elif Gürsoy et al.
European journal of medicinal chemistry, 42(3), 320-326 (2006-12-06)
A series of arylidenehydrazides (3a-3i) were synthesized from [6-(4-bromophenyl)imidazo[2,1-b]thiazol-3-yl]acetic acid hydrazide. The structures of new compounds were determined by analytical and spectral (IR, (1)H NMR, (13)C NMR, EIMS) methods. The synthesized compounds (3a-3i) were evaluated in the National Cancer Institute's
Jignesh M Doshi et al.
Journal of medicinal chemistry, 49(26), 7731-7739 (2006-12-22)
The structure-activity relationship studies of ethyl 2-amino-6-cyclopentyl-4-(1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate (1, HA 14-1), an antagonist of the antiapoptotic Bcl-2 proteins, are reported. A series of analogues of 1 with varied functional groups at the 6-position of the chromene ring were synthesized. These candidates
Gustavo H B Maegawa et al.
The Journal of biological chemistry, 282(12), 9150-9161 (2007-01-24)
Late-onset GM2 gangliosidosis is composed of two related, autosomal recessive, neurodegenerative diseases, both resulting from deficiency of lysosomal, heterodimeric beta-hexosaminidase A (Hex A, alphabeta). Pharmacological chaperones (PC) are small molecules that can stabilize the conformation of a mutant protein, allowing
Alex M Aronov et al.
Journal of medicinal chemistry, 51(5), 1214-1222 (2008-02-22)
Small molecule protein kinase inhibitors are widely employed as biological reagents and as leads in the design of drugs for a variety of diseases. We investigated the phenomenon of kinase-likeness, i.e., the propensity of ligands to inhibit protein kinases, in
Lucie Brulíková et al.
European journal of medicinal chemistry, 45(9), 3588-3594 (2010-07-08)
A series of uridine analogues modified at the 5-position with the 5-[alkoxy-(4-nitrophenyl)-methyl] moiety was synthesized. Nucleosides were formed as a mixture of two diastereoisomers, which were separated and tested for their cytotoxic activity in vitro against different cancer cell lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service