Skip to Content
Merck
All Photos(1)

Documents

606448

Sigma-Aldrich

Acrolein-13C3

≥99 atom % 13C, ≥90% (CP), contains hydroquinone as stabilizer

Synonym(s):

2-Propenal-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H213C=13CH13CHO
Molecular Weight:
59.04
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥90% (CP)

contains

hydroquinone as stabilizer

impurities

≤10% water

bp

53 °C (lit.)

mp

-87 °C (lit.)

density

0.883 g/mL at 25 °C

mass shift

M+3

storage temp.

−20°C

SMILES string

[13CH2]=[13CH][13CH]=O

InChI

1S/C3H4O/c1-2-3-4/h2-3H,1H2/i1+1,2+1,3+1

InChI key

HGINCPLSRVDWNT-VMIGTVKRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service