Skip to Content
Merck
All Photos(1)

Key Documents

563250

Sigma-Aldrich

4-Methyl-1-piperazinecarbonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H8N2CH3COCl
CAS Number:
Molecular Weight:
162.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4980 (lit.)

bp

278-279 °C (lit.)

density

1.200 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C(Cl)=O

InChI

1S/C6H11ClN2O/c1-8-2-4-9(5-3-8)6(7)10/h2-5H2,1H3

InChI key

FBAIGEMWTOSCRU-UHFFFAOYSA-N

General description

4-Methyl-1-piperazinecarbonyl chloride can be synthesized from 1-methylpiperazine by reacting with phosgene.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. J. Brown et al.
The Chemistry of Heterocyclic Compounds, The Pyrazines: Supplement I, 58 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service