Skip to Content
Merck
All Photos(1)

Key Documents

556335

Sigma-Aldrich

2-Allyloxytetrahydropyran

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

contains

~1% K2CO3 as stabilizer

refractive index

n20/D 1.4275 (lit.)

bp

76-77 °C/25 mmHg (lit.)

density

0.940 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCOC1CCCCO1

InChI

1S/C8H14O2/c1-2-6-9-8-5-3-4-7-10-8/h2,8H,1,3-7H2

InChI key

NRAKYXXDEXCVMV-UHFFFAOYSA-N

General description

2-Allyloxytetrahydropyran can undergo Heck coupling reaction with various aryl bromides in the presence of [PdCl(C3H5)]2/cis,cis,cis-1,2,3,4-tetrakis(diphenylphosphan-ylmethyl)cyclopentane [catalyst].

Caution

Refrigerate

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heck reaction of protected allyl alcohols with aryl bromides catalyzed by a tetraphosphanepalladium complex.
Berthiol F, et al.
European Journal of Organic Chemistry, 2005(7), 1367-1377 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service