Skip to Content
Merck
All Photos(2)

Documents

476854

Sigma-Aldrich

Chlorodiethylisopropylsilane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHSi(C2H5)2Cl
CAS Number:
Molecular Weight:
164.75
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.438 (lit.)

bp

73 °C/32 mmHg (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

CC[Si](Cl)(CC)C(C)C

InChI

1S/C7H17ClSi/c1-5-9(8,6-2)7(3)4/h7H,5-6H2,1-4H3

InChI key

OSBPGFIPLLCQMO-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - Water-react 3

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

100.0 °F - closed cup

Flash Point(C)

37.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhixiong Liu et al.
Neuron, 102(3), 587-601 (2019-03-17)
Nucleoporins (Nups) are involved in neural development, and alterations in Nup genes are linked to human neurological diseases. However, physiological functions of specific Nups and the underlying mechanisms involved in these processes remain elusive. Here, we show that tissue-specific depletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service