Skip to Content
Merck
All Photos(2)

Documents

397172

Sigma-Aldrich

(S)-(+)-2-Methylpiperazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D +6.8°, c = 1 in ethanol

mp

91-93 °C (lit.)

SMILES string

C[C@H]1CNCCN1

InChI

1S/C5H12N2/c1-5-4-6-2-3-7-5/h5-7H,2-4H2,1H3/t5-/m0/s1

InChI key

JOMNTHCQHJPVAZ-YFKPBYRVSA-N

Application

Valuable synthon for quinoline antibacterials.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic preparations and procedures international, 22, 761-761 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service