348228
3-(Trifluoromethyl)phenylhydrazine
technical grade, 90%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CF3C6H4NHNH2
CAS Number:
Molecular Weight:
176.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
technical grade
Assay
90%
refractive index
n20/D 1.504 (lit.)
bp
80-83 °C/9 mmHg (lit.)
density
1.348 g/mL at 25 °C (lit.)
SMILES string
NNc1cccc(c1)C(F)(F)F
InChI
1S/C7H7F3N2/c8-7(9,10)5-2-1-3-6(4-5)12-11/h1-4,12H,11H2
InChI key
RSESUCWJKLHXEZ-UHFFFAOYSA-N
General description
3-(Trifluoromethyl)phenylhydrazine can be prepared by reduction of 3-(trifluoromethyl)phenyldiazonium chloride.
Application
3-(Trifluoromethyl)phenylhydrazine may be used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides, having antitubercular and fungicidal activity.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
224.6 °F - closed cup
Flash Point(C)
107 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of new fluorine substituted hydrazinecarboxamides and hydrazine carbothioamides having antitubercular and fungicidal activity
Nalavde YM and Joshi V.
Indian J. Chem. B, 39(8), 634-637 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service